Skip to Content
MilliporeSigma

04-1583

Anti-Interleukin 21 (IL-21) Antibody, clone 1G8

clone AB43.50.32H5.1G8, from mouse

Synonym(s):

interleukin 2, interleukin-2, interleukin-21 isoform

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
AB43.50.32H5.1G8, monoclonal
Application:
ELISA, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AB43.50.32H5.1G8, monoclonal

species reactivity

human

technique(s)

ELISA: suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IL21(59067)

General description

Interleukin 21 (IL-21) is a cytokine that has immunoregulatory activity on cells of the immune system, including natural killer (NK) cells and cytotoxic T cells that can destroy virally infected or cancerous cells. IL-21 induces cell division/proliferation in its target cells and is expressed in activated human CD4+ T cells. IL-21 may promote the transition between innate and adaptive immunity and may play a role in proliferation and maturation of natural killer (NK) cells in synergy with IL15. IL-21 expression is up-regulated in Th2 and Th17 subsets of T helper cells, as well as follicular T cells. Furthermore IL-21 is expressed in NK T cells regulating the function of these cells. IL-21 is also produced by Hodgkin′s lymphoma (HL) cancer cells and may explain a great deal of the behavior of classical Hodgkin′s lymphoma including clusters of other immune cells gathered around HL cells in cultures.
Two isoforms are observed at ~16 and ~18 kDa

Immunogen

Full length recombinant protein corresponding to human IL-21.

Application

ELISA Analysis: A previous lot was used by an independent laboratory in ELISA. (Dr. Gerard Zurawski, Baylor Institute for Immunology Research, Dallas, TX)
This Anti-Interleukin 21 (IL-21) Antibody, clone 1G8 is validated for use in WB, ELISA for the detection of Interleukin 21 (IL-21).

Physical form

Format: Purified

Analysis Note

Control
Human IL-21 recombinant protein
Evaluated by Western Blot from human IL-21 recombinant protein.

Western Blot Analysis: 0.5 µg/mL of this antibody detected IL-21 on 10 µg of human IL-21 recombinant protein.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library