Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
2EI-7, monoclonal
Application:
FACS, WB
Citations:
38
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2EI-7, monoclonal
species reactivity
mouse
species reactivity (predicted by homology)
human (based on 100% sequence homology)
technique(s)
flow cytometry: suitable, western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PYCARD(29108)
Related Categories
General description
Apoptosis is regulated by death domain (DD) and/or caspase recruitment domain (CARD) containing molecules and a caspase family of proteases. CARD containing cell death regulators include RAIDD, RICK, Bcl10, Apaf-1, ARC, caspase-2 and caspase-9. A novel CARD domain containing protein has been identified in human and mouse and designated ASC and TMS1. Irregular expression of ASC/TMS1 induced apoptosis through activation of caspase-9 and inhibited the survival of human breast cancer cells. Overexpression of ASC/TMS1 induced DNA fragmentation. ASC/TMS1 is expressed in a variety of human and mouse tissues.
~22 kDa
Immunogen
Epitope: Unknown
Full length recombinant human ASC.
Application
Detect ASC using this Anti-ASC Antibody, clone 2EI-7 validated for use in WB, FC.
Research Category
Inflammation & Immunology
Inflammation & Immunology
Research Sub Category
Apoptosis - Additional
Apoptosis - Additional
Biochem/physiol Actions
This antibody recognizes ASC.
Physical form
Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Control
Mouse spleen tissue lysate
Mouse spleen tissue lysate
Evaluated by Western Blot on mouse spleen tissue lysate.
Western Blot Analysis: 0.5 µg/mL of this antibody detected ASC in mouse spleen tissue lysate.
Western Blot Analysis: 0.5 µg/mL of this antibody detected ASC in mouse spleen tissue lysate.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sonal Khare et al.
Nature immunology, 15(4), 343-353 (2014-02-18)
The innate immune system responds to infection and tissue damage by activating cytosolic sensory complexes called 'inflammasomes'. Cytosolic DNA is sensed by AIM2-like receptors (ALRs) during bacterial and viral infections and in autoimmune diseases. Subsequently, recruitment of the inflammasome adaptor
Wei Zhu et al.
Neuroscience, 343, 77-84 (2016-12-08)
Lipopolysaccharide (LPS) might affect the central nervous system by causing neuroinflammation, which subsequently leads to brain damage and dysfunction. In this study, we evaluated the role of nod-like receptor pyrin domain-containing protein 3 (NLRP3) inflammasome activation in long-term behavioral alterations
Chunqing Guo et al.
ACS chemical neuroscience, 8(10), 2194-2201 (2017-06-28)
In our efforts to develop novel small-molecule inhibitors for the NOD-like receptor family pyrin-domain-containing 3 (NLRP3) inflammasome as potential disease-modifying agents to treat neurological disorders including multiple sclerosis (MS), a hydroxyl sulfonamide analogue JC-171 has been rationally designed and biologically
Si Ming Man et al.
Journal of immunology (Baltimore, Md. : 1950), 191(10), 5239-5246 (2013-10-15)
Nucleotide-binding oligomerization domain-like receptors (NLRs) detect pathogens and danger-associated signals within the cell. Salmonella enterica serovar Typhimurium, an intracellular pathogen, activates caspase-1 required for the processing of the proinflammatory cytokines, pro-IL-1β and pro-IL-18, and pyroptosis. In this study, we show
Chromatin dynamics during DSB repair.
Martin Falk,Emilie Lukasova,Barbora Gabrielova,Vladan Ondrej,Stanislav Kozubek
Biochimica et Biophysica Acta null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service