Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04-080

Sigma-Aldrich

Anti-BAF250a/ARID1a Antibody, clone PSG3

clone PSG3, Upstate®, from mouse

Synonym(s):

Anti-B120, Anti-BAF250, Anti-BAF250a, Anti-BM029, Anti-C1orf4, Anti-CSS2, Anti-ELD, Anti-MRD14, Anti-OSA1, Anti-P270, Anti-SMARCF1, Anti-hELD, Anti-hOSA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PSG3, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ARID1A (8289)

Specificity

BAF250a/ARID1a

Immunogen

GST fusion protein corresponding to 419 residues in the mid-portion of human BAF250a/ARID1a

Application

Anti-BAF250a/ARID1a Antibody, clone PSG3 is an antibody against BAF250a/ARID1a for use in WB, ChIP, IC & IHC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Transcription Factors

Quality

Routinely evaluated by immunoblot.

Target description

~270 kDa

Physical form

100 μg of Protein A purified mouse monoclonal IgG in 100μl of 1X PBS, pH 7.0, 0.1% Azide.
Format: Purified
Protein A purified

Storage and Stability

2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coexistent ARID1A-PIK3CA mutations promote ovarian clear-cell tumorigenesis through pro-tumorigenic inflammatory cytokine signalling.
Chandler, RL; Damrauer, JS; Raab, JR; Schisler, JC; Wilkerson, MD; Didion, JP; Starmer et al.
Nature Communications null
Hiroko Inoue et al.
The Journal of biological chemistry, 277(44), 41674-41685 (2002-08-30)
The mammalian SWI/SNF-related complexes facilitate gene transcription by remodeling chromatin using the energy of ATP hydrolysis. The recruitment of these complexes to promoters remains poorly understood and may involve histone modifications or direct interactions with site-specific transcription factors or other
Zuqin Nie et al.
Molecular and cellular biology, 23(8), 2942-2952 (2003-04-01)
The SWI/SNF family of chromatin-remodeling complexes has been discovered in many species and has been shown to regulate gene expression by assisting transcriptional machinery to gain access to their sites in chromatin. Several complexes of this family have been reported
Essential role of ARID2 protein-containing SWI/SNF complex in tissue-specific gene expression.
Xu, F; Flowers, S; Moran, E
The Journal of Biological Chemistry null
Radhika Mathur et al.
Nature genetics, 49(2), 296-302 (2016-12-13)
Genes encoding subunits of SWI/SNF (BAF) chromatin-remodeling complexes are collectively mutated in ∼20% of all human cancers. Although ARID1A is the most frequent target of mutations, the mechanism by which its inactivation promotes tumorigenesis is unclear. Here we demonstrate that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service