Skip to Content
MilliporeSigma

03-34-0001

H-Gly-Pro-Arg-Pro-OH

Inhibitor of fibrinogen aggregation and fibrin polymerization.

Synonym(s):

H-Gly-Pro-Arg-Pro-OH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H31N7O5
CAS Number:
Molecular Weight:
425.48
UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

technique(s)

protein extraction: suitable

solubility

5% acetic acid: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N2([C@@H](CCC2)C(=O)O)C(=O)[C@@H](NC(=O)[C@H]1N(CCC1)C(=O)CN)CCCN=C(N)N

InChI

1S/C18H31N7O5/c19-10-14(26)24-8-2-5-12(24)15(27)23-11(4-1-7-22-18(20)21)16(28)25-9-3-6-13(25)17(29)30/h11-13H,1-10,19H2,(H,23,27)(H,29,30)(H4,20,21,22)/t11-,12-,13-/m0/s1

InChI key

WXPZDDCNKXMOMC-AVGNSLFASA-N

General description

Inhibitor of fibrinogen aggregation and fibrin polymerization.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Other Notes

Barnard, M.R., et al. 1999. Transfusion39, 73.
Rijkers, D.T., et al. 1998. Thromb. Res. 89, 161.
Achyuthan, K.E., and Dobson, J.V. 1986. Biochim. Biophys. Acta872, 261.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
03-34-0001-25MG04055977206784