Skip to Content
MilliporeSigma

03-0206-00

SMC® Human SNAP-25 High Sensitivity Immunoassay Kit

Used for ultrasensitive quantification of Human SNAP-25 in human serum, plasma, and cerebrospinal fluid (CSF)

Synonym(s):

SNAP-25 Single Molecule Counting kit, SNAP-25 immunoassay kit, SNAP-25 ultra-sensitive immunoassay, Synaptosomal-associated protein 25 immunoassay kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.77
UNSPSC Code:
12161503
Species reactivity:
human
Technique(s):
single molecule counting: suitable
Usage:
 kit sufficient for 96 wells
Compatibility:
for use with FemtoQuest and SMCxPRO® instruments
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

SMC® Human SNAP-25 High Sensitivity Immunoassay Kit,

Quality Segment

description

Used for ultrasensitive quantification of Human SNAP-25 in human serum, plasma, and cerebrospinal fluid (CSF)

product line

SMC®

usage

 kit sufficient for 96 wells

species reactivity

human

technique(s)

single molecule counting: suitable

input

serum
plasma (K2 EDTA)
cerebrospinal fluid (CSF)

UniProt accession no.

compatibility

for use with FemtoQuest and SMCxPRO® instruments

shipped in

wet ice

storage temp.

2-8°C

General description

Synaptosomal-associated protein 25 (SNAP-25) plays a crucial role in synaptic transmission in the nervous system. As a component of the SNARE complex, it facilitates the fusion of synaptic vesicles with the presynaptic membrane, enabling the release of neurotransmitters into the synaptic cleft. By promoting efficient neurotransmitter release, SNAP-25 is essential for proper neuronal communication and signaling.

SNAP-25 has been implicated in various conditions, including Alzheimer′s disease, schizophrenia, and multiple sclerosis. Abnormalities in SNAP-25 expression or function can disrupt synaptic transmission, leading to impaired neuronal communication and contributing to cognitive decline and other symptoms associated with these disorders. Alterations in SNAP-25 levels often correlate with disease severity, highlighting its importance in neuroinflammation and synaptic dysfunction. Understanding the role of SNAP-25 in these processes can help identify potential therapeutic targets and inform strategies aimed at restoring synaptic function and improving outcomes for neurological disorders.

Application

Alzheimer′s Disease / Aging / Nervous System / Neurodegenerative Diseases / Neurofilament & Neuron Metabolism / Neuronal & Glial Markers / Neurodegenerative Medicine / Neuroscience

Features and Benefits

  • Ultra-Sensitive Measurement: Accurately detects SNAP-25 at femtogram per milliliter levels in serum, plasma and CSF.
  • Reliable Data: An improved signal-to-noise ratio extends the dynamic range to guarantee precise and trustworthy assay results.
  • Easy to Adopt: Follow a well-known sandwich immunoassay protocol to seamlessly incorporate SMC®technology into research processes.
  • Automated Workflow: Designed for compatibility with automated liquid handling systems, this assay minimizes manual tasks and enhances laboratory efficiency.
  • Expert Support: Get technical assistance from specialized field scientists and technical teams.

Packaging

96 well assay

Analysis Note

  • Sensitivity: LLOQ 260 fg/mL* (SMCxPRO® Instrument)
  • Standard Curve Range: 130-300,000 fg/mL
  • Intra-assay CV: <15%
  • Inter-assay CV: <20%
  • Linearity: Serum: 112% Plasma: 110% CSF: 97%
  • Recovery: Serm: 82% Plasma: 91% CSF: 89%
*Indicative LLOQ created from average of 14 independent in house runs, where >80% met same value defined with CV & Recovery +/- 20%. Refer to CoA for lot specific performance.

Other Notes

Although SNAP-25 was detectable in non-human primate (rhesus macaque and cynomolgus monkey), feline, rat, mouse, canine samples, the assay has not been verified for these species.

Legal Information

FEMTOQUEST is a trademark of Merck KGaA, Darmstadt, Germany
SMC is a registered trademark of Singulex
SMCxPRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - ED ENV 1 - Eye Irrit. 2 - Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library