Skip to Content
MilliporeSigma

01-201

Heregulin-β3, Epidermal growth factor domain

liquid, suitable for cell culture

Synonym(s):

EGF Domain

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Heregulin-β3, Epidermal growth factor (EGF) domain, The Heregulin-β3 protein, Epidermal growth factor (EGF) domain is available in a 100 µg format.

Quality Segment

form

liquid

manufacturer/tradename

Upstate®

technique(s)

cell culture | mammalian: suitable

input

sample type mesenchymal stem cell(s)
sample type cardiac stem cell(s)

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

Gene Information

human ... NRG1(3084)

General description

Product Source: Thrombin cleaved GST-fusion protein corresponding to the EGF domain (residues 178-241) of heregulin-beta 3.

Physical form

PBS with 30% glycerol. Liquid.

Analysis Note

Routinely evaluated by Receptor Activation Analysis.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library