Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T-046

Supelco

Testosterone-d3 (16, 16, 17-d3) solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Testosterone-d3 solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[H]C1([H])C[C@]2([H])[C@@]3([H])CCC4=CC(=O)CC[C@]4(C)[C@]3([H])CC[C@@]2(C)[C@]1([H])O

InChI

1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1/i6D2,17D

InChI key

MUMGGOZAMZWBJJ-HZRGXLBSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard for testosterone testing or isotope dilution methods by GC/MS or LC/MS for sports testing, urine drug testing or diagnostic testing by LC-MS/MS or GC/MS. Testosterone, a hormone from the androgen group, is an important sex hormone and anabolic steroid used as a performance enhancing drug.

Application


  • First detection/quantification of roxadustat in hair with a new liquid chromatography with tandem mass spectrometry method: This study highlights the advanced LC-MS/MS methodologies that can be adapted for the detection and quantification of deuterated compounds like Testosterone-d₃ in biological samples, providing precision in pharmacokinetic studies (Alvarez et al., 2023).

  • A quantitative method for the simultaneous detection of SR9009 and SR9011 in equine plasma: Demonstrates the application of liquid chromatography-electrospray ionization-tandem mass spectrometry, a technique that can be used for accurately measuring stable isotope-labeled compounds such as Testosterone-d₃ in complex matrices (Kwak et al., 2022).

  • Collision energy-breakdown curves - An additional tool to characterize MS/MS methods: Discusses the optimization of mass spectrometry parameters which can enhance the detection of deuterated drugs like Testosterone-d₃, crucial for detailed metabolic studies (Mörlein et al., 2020).

  • Detection and quantification of 12 anabolic steroids and analogs in human whole blood and 20 in hair using LC-HRMS/MS: Explores high-resolution mass spectrometry for detecting substances in biological samples, relevant for the precise analysis of Testosterone-d₃ in doping control and clinical research (Fabresse et al., 2017).

  • Determination of anabolic agents in dietary supplements by liquid chromatography-high-resolution mass spectrometry: Focuses on the reliable identification of performance-enhancing drugs, which can include the application of Testosterone-d₃ as an internal standard to ensure accuracy and traceability in analytical testing (Odoardi et al., 2015).



Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.

Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.

Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.

Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service