Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Q-002

Supelco

Quetiapine-D8 hemifumarate solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
(C21D8H17N3O2S)2 · C4H4O4
CAS Number:
Molecular Weight:
899.18
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OC(=O)\C=C\C(O)=O.[2H]C1([2H])N(CCOCCO)C([2H])([2H])C([2H])([2H])N(C2=Nc3ccccc3Sc4ccccc24)C1([2H])[2H].[2H]C5([2H])N(CCOCCO)C([2H])([2H])C([2H])([2H])N(C6=Nc7ccccc7Sc8ccccc68)C5([2H])[2H]

InChI

1S/2C21H25N3O2S.C4H4O4/c2*25-14-16-26-15-13-23-9-11-24(12-10-23)21-17-5-1-3-7-19(17)27-20-8-4-2-6-18(20)22-21;5-3(6)1-2-4(7)8/h2*1-8,25H,9-16H2;1-2H,(H,5,6)(H,7,8)/b;;2-1+/i2*9D2,10D2,11D2,12D2;

InChI key

ZTHJULTYCAQOIJ-UXUYVNHUSA-N

General description

Quetiapine, marketed as Seroquel®, is an atypical antipsychotic drug for the treatment of depression, bipolar disorder, and schizophrenia. This new internal standard is applicable to numerous GCMS and LCMS applications including quantitation of patient levels, forensic and forensic toxicology analysis, isotope dilution methods, and calibrator preparation.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Seroquel is a registered trademark of Astrazeneca UK
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service