Skip to Content
MilliporeSigma

P-044

Promethazine hydrochloride

vial of 25 mg, certified reference material, Cerilliant®

Synonym(s):

10-[2-(Dimethylamino)propyl]phenothiazine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H20N2S · HCl
CAS Number:
Molecular Weight:
320.88
EC Number:
200-375-2
UNSPSC Code:
41116107
PubChem Substance ID:
Beilstein/REAXYS Number:
4166397
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

solid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

vial of 25 mg

manufacturer/tradename

Cerilliant®

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

room temp

SMILES string

Cl[H].CC(CN1c2ccccc2Sc3ccccc13)N(C)C

InChI

1S/C17H20N2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H

InChI key

XXPDBLUZJRXNNZ-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

General description

Promethazine is a phenothiazine analog prescribed for treatment of motion sickness, nausea, vomiting and allergies. This Certified Reference Material is suitable for GC/MS or LC/MS methods in promethazine testing for forensic analysis, urine drug testing, or clinical toxicology. Sold under the trade names Phenergan® and Phenadoz®, promethazine has antihistamine, sedative, and mild antipsychotic effects. The compound has also been been found in the recreational drug "Purple Drank."

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Phenadoz is a registered trademark of Watson Laboratories, Inc.
Phenergan is a registered trademark of Delcor Asset Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Promethazine hydrochloride


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library