Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

O-057

Supelco

Oxazepam glucuronide lithium salt solution

100 μg/mL in methanol ((as free carboxylate)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18ClLiN2O8
Molecular Weight:
468.77
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

description

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol ((as free carboxylate))

format

single component solution

storage temp.

-10 to -25°C

General description

Oxazepam glucuronide is a major urinary metabolite of the benzodiazepine drug, oxazepam. Oxazepam, marketed as Alepam, Medopam, or Murelax, has been used extensively since the 1960s for treatment of anxiety and insomnia and in the control of symptoms of alcohol withdrawal. This certified reference solution is suitable for GC/MS or LC/MS applications with oxazepam or oxazepam glucuronide such as urine drug testing, pain prescription monitoring, toxicology analysis, or forensic testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service