Skip to Content
MilliporeSigma

O-030

Oxymorphone-3-β-D-glucuronide solution

100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H27NO10
CAS Number:
Molecular Weight:
477.46
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol: water (1:1)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CN1CC[C@]23[C@H]4Oc5c(O[C@@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(O)=O)ccc(C[C@@H]1[C@]2(O)CCC4=O)c35

InChI

1S/C23H27NO10/c1-24-7-6-22-13-9-2-3-11(32-21-16(28)14(26)15(27)18(34-21)20(29)30)17(13)33-19(22)10(25)4-5-23(22,31)12(24)8-9/h2-3,12,14-16,18-19,21,26-28,31H,4-8H2,1H3,(H,29,30)/t12-,14+,15+,16-,18+,19+,21-,22+,23-/m1/s1

InChI key

ILRLBQVIWURYLX-WUDROUKASA-N

General description

A certified Snap-N-Spike® solution of Oxymorphone-3ß-D-glucuronide suitable for use in a variety of GC/MS or LC/MS applications such as pain prescription monitoring, clinical toxicology, pharmaceutical research, forensic analysis, or urine drug testing. Oxymorphone-3ß-D-glucuronide is a major metabolite of oxymorphone in urine and plasma. Oxymorphone, sold under trade names such as Opana®, Numorphan®, and Numorphone, is an opiate analgesic indicated for relief of moderate to severe pain.

Legal Information

Opana is a registered trademark of Endo Pharmaceuticals Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Numorphan is a registered trademark of Endo Pharmaceuticals Inc.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

75.2 °F - closed cup

flash_point_c

24 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library