Skip to Content
MilliporeSigma
All Photos(1)

Documents

N-049

Supelco

(±)-Norsertraline hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16Cl3N
CAS Number:
Molecular Weight:
328.66
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

N[C@H]1CC[C@@H](C2=CC(Cl)=C(Cl)C=C2)C3=C1C=CC=C3.[H]Cl

InChI

1S/C16H15Cl2N.ClH/c17-14-7-5-10(9-15(14)18)11-6-8-16(19)13-4-2-1-3-12(11)13;/h1-5,7,9,11,16H,6,8,19H2;1H/t11-,16-;/m0./s1

InChI key

YKXHIERZIRLOLD-RISSCEEYSA-N

General description

Norsertraline is a major urinary metabolite and N-desmethyl analog of the SSRI antidepressant sertraline. Marketed under trade names such as Zoloft® and Lustral, sertraline is used to treat major depressive, obsessive-compulsive and panic and social anxiety disorders in both adults and children.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Zoloft is a registered trademark of Pfizer, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramagiri Suma et al.
Journal of analytical toxicology, 30(6), 395-399 (2006-07-29)
Zoloft (sertraline hydrochloride) is one of the antidepressant medications used to treat depression, obsessive-compulsive disorder, and social anxiety disorder. The practice of embalming a cadaver is common, yet it may create problems for forensic toxicologists if the case was not
Alessandro Musenga et al.
Electrophoresis, 28(11), 1823-1831 (2007-04-24)
A method has been developed for the analysis of the antidepressant drug sertraline together with its main metabolite N-desmethylsertraline (DMS) in human plasma. It is based on CE with LIF detection (lambda = 488 nm). A SPE procedure is employed
L M Hesse et al.
Drug metabolism and disposition: the biological fate of chemicals, 28(10), 1176-1183 (2000-09-21)
The in vitro biotransformation of bupropion to hydroxybupropion was studied in human liver microsomes and microsomes containing heterologously expressed human cytochromes P450 (CYP). The mean (+/-S.E.) K(m) in four human liver microsomes was 89 (+/-14) microM. In microsomes containing cDNA-expressed
V L Serebruany et al.
Pharmacological research, 43(5), 453-462 (2001-06-08)
Recently, clinical depression has been identified as an independent risk factor for increased mortality in patients following acute coronary events. Although the underlying mechanisms of this link remain uncertain, increased platelet activity has been suggested but never proven as the
Bryan W Brooks et al.
Environmental toxicology and chemistry, 24(2), 464-469 (2005-02-22)
Increasing evidence indicates widespread occurrence of pharmaceuticals and personal care products (PPCPs) in municipal effluent discharges and surface waters. Studies that characterize the fate and effects of PPCPs in aquatic systems are limited, and to our knowledge, data regarding pharmaceutical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service