Skip to Content
MilliporeSigma

N-045

Norbuprenorphine glucuronide solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Norbuprenorphine glucuronide solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C31H43NO10
CAS Number:
Molecular Weight:
589.67
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-659-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CO[C@]12CC[C@@]3(C[C@@H]1[C@](C)(O)C(C)(C)C)[C@H]4Cc5ccc(O[C@@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(O)=O)c7O[C@@H]2[C@]3(CCN4)c57

InChI

1S/C31H43NO10/c1-27(2,3)28(4,38)16-13-29-8-9-31(16,39-5)26-30(29)10-11-32-17(29)12-14-6-7-15(22(42-26)18(14)30)40-25-21(35)19(33)20(34)23(41-25)24(36)37/h6-7,16-17,19-21,23,25-26,32-35,38H,8-13H2,1-5H3,(H,36,37)/t16-,17-,19+,20+,21-,23+,25-,26-,28+,29-,30+,31-/m1/s1

InChI key

OOLGGESLZHGXGC-TXYLTYGDSA-N

General description

This analytical reference standard is applicable for use in LC/MS or GC/MS applications for pain prescription monitoring, urine drug testing, clinical toxicology, or forensic analysis. Norbuprenorphine glucuronide is a urinary metabolite of the semi-synthetic opioid analgesic buprenorphine. Buprenorphine, sold under the trade names Temgesic and Subutex®, is prescribed for the treatment of chronic pain and heroin addiction.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Subutex is a registered trademark of RB Pharmaceuticals Limited

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library