Skip to Content
MilliporeSigma

M-262

(±)-Metanephrine hydrochloride solution

1 mg/mL in methanol ((as free base)), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Metanephrine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H15NO3 · HCl
CAS Number:
Molecular Weight:
233.69
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol ((as free base))

format

single component solution

storage temp.

-10 to -25°C

SMILES string

[Cl-].N(CC(O)c1cc(c(cc1)O)OC)C.[H+]

InChI key

HRIQFVCFOPJYEQ-UHFFFAOYSA-N

General description

Measurement of metanephrine, a metabolite of epinephrine, is a useful tool in the diagnosis of diseases such as pheochromocytoma and adrenal medullary neoplasm. This Certified Reference Material is suitable for LC-MS/MS monitoring of metanephrine levels in plasma or urine for diagnostic testing, endocrinology, or clinical chemistry.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library