Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C1213CH13D3N2 · HCl
CAS Number:
Molecular Weight:
240.75
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
grade
certified reference material
Quality Segment
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol ((as free base))
format
single component solution
storage temp.
-10 to -25°C
General description
A stable-labled internal standard suitable for quantitation of Dexmedetomidine levels in urine, serum, or plasma by LC/MS or GC/MS for therapeutic drug monitoring, pharmaceutical research, forensic analysis, clinical toxicology and urine drug testing. Dexmedetomidine, sold under brand names such as Precedex, Dexdor, Dexdomitor or Sileo, is an anxiety reducing, sedative, and pain medication.
Snap-N-Spike®/Snap-N-Shoot®
Legal Information
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
WGK
WGK 2
Flash Point (°F)
49.5 °F
Flash Point (°C)
9.7 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


