Skip to Content
MilliporeSigma
All Photos(1)

Documents

M-185

Supelco

(±)-threo-Methylphenidate-D4 hydrochloride solution

100 μg/mL in methanol (as free base), certified reference material, Cerilliant®

Synonym(s):

Deuterated methylphenidate hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15D4NO2 · HCl
Molecular Weight:
273.79
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(OC)[C@H]([C@]1([H])CC([2H])([2H])C([2H])([2H])CN1)C2=CC=CC=C2.[H]Cl

InChI

1S/C14H19NO2.ClH/c1-17-14(16)13(11-7-3-2-4-8-11)12-9-5-6-10-15-12;/h2-4,7-8,12-13,15H,5-6,9-10H2,1H3;1H/t12-,13?;/m0./s1/i5D2,6D2;

InChI key

JUMYIBMBTDDLNG-HGUJNOQNSA-N

General description

This internal standard is suitable for quantitation of methylphenidate levels in urine, serum or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology or forensic analysis. Methylphenidate is a psychostimulant . The drug is prescribed for the treatment of attention-deficit hyperactivity disorder (ADHD), tachycardia and narcolepsy. 

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Thomele et al.
Crystal growth & design, 21(8), 4674-4682 (2021-08-13)
Developing simple, inexpensive, and environmentally benign approaches to integrate morphologically well-defined nanoscale building blocks into larger high surface area materials is a key challenge in materials design and processing. In this work, we investigate the fundamental surface phenomena between MgO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service