Skip to Content
MilliporeSigma

M-160

(±)-4-Methylephedrine-D3 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Deuterated 4-methylephedrine hydrochloride solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H14D3NO · HCl
Molecular Weight:
218.74
NACRES:
NA.24
UNSPSC Code:
41116107
EC Number:
200-659-6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CC(NC([2H])([2H])[2H])C(O)C1=CC=C(C)C=C1.Cl

InChI

1S/C11H17NO.ClH/c1-8-4-6-10(7-5-8)11(13)9(2)12-3;/h4-7,9,11-13H,1-3H3;1H/i3D3;

InChI key

DDFKODAFZRBBOK-FJCVKDQNSA-N

General description

A bath salt cathinone analog and primary urinary metabolite of mephedrone. Mephedrone, also known as 4-methylmethcathinone, is a synthetic cathinone and stimulant widely found in numerous bath salt blends across the US, Europe, and Asia. This certified Snap-N-Spike® solution is suitable as an internal standard for quantitation of (±)-4-Methylephedrine by LC/MS or GC/MS for applications in urine drug testing, clinical toxicology, or forensic testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F

flash_point_c

9.7 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library