Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M-146

Supelco

3,4-Methylenedioxypyrovalerone HCl (MDPV) solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

3,4-Methylenedioxypyrovalerone hydrochloride, MDPV hydrochloride, MDPV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO3 · HCl
CAS Number:
Molecular Weight:
311.80
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.N3(CCCC3)C(CCC)C(=O)c1cc2c(cc1)OCO2

InChI

1S/C16H21NO3.ClH/c1-2-5-13(17-8-3-4-9-17)16(18)12-6-7-14-15(10-12)20-11-19-14;/h6-7,10,13H,2-5,8-9,11H2,1H3;1H

InChI key

PYQZNWFQAMFAMT-UHFFFAOYSA-N

General description

3,4-Methylenedioxypyrovalerone, also known as MDPV, Mtv, or Super Coke, is a new designer drug and stimulant with cases of abuse reported in Europe and the US. In mid- to late-2010, MDPV started appearing in the US as a legal drug alternative marketed as “bath salts” under names including Ivory Wave, Tranquility, and Cloud 9. This certified solution standard offers numerous applications in GC/MS or LC/MS testing from forensic analysis and clinical toxicology to urine drug monitoring.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This article highlights the impact that sample matrix effects can have on LC/MS response and discusses two novel approaches to reduce it.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service