Skip to Content
MilliporeSigma
All Photos(1)

Documents

M-078

Supelco

2-Mononitroglycerin solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO5
CAS Number:
Molecular Weight:
137.09
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

OCC(CO)O[N+]([O-])=O

InChI

1S/C3H7NO5/c5-1-3(2-6)9-4(7)8/h3,5-6H,1-2H2

InChI key

QYZBMMOLPVKAPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Mononitroglycerin is a major plasma metabolite of the pharmaceutical vasodilator nitroglycerin. This Certified Spiking Solution® is suitable for use in chromatography or mass spectrometry-based applications in pharmaceutical research or clinical toxicology. Nitroglycerin is used to treat heart conditions such as angina and chronic heart failure.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Ducrocq et al.
Biotechnology and applied biochemistry, 12(3), 325-330 (1990-06-01)
Various microorganisms have been evaluated for their ability to hydrolyze glyceryl trinitrate (GTN) to glyceryl dinitrates and mononitrates. Provided that the GTN extracellular concentration was under the lethal dose, metabolite formation and regioselectivity depend on the nature of the strain
M Leitold et al.
Arzneimittel-Forschung, 36(12), 1752-1756 (1986-12-01)
Comparative Hemodynamic Activity of Glyceryl 2-Nitrate and Glyceryl Trinitrate in Various Species and Pharmacokinetics of Glyceryl 2-Nitrate in Dog A single dose of glyceryl 2-nitrate (G-2-N) had practically identical hypotensive and cardiovascular activity when administered intravenously or enterally to the
M Leitold et al.
Arzneimittel-Forschung, 37(6), 692-698 (1987-06-01)
Glyceryl 2-nitrate (G-2-N), which is the major metabolite of glyceryl trinitrate (GTN, Nitro Mack, glyceryl 1-nitrate (G-1-N) and isosorbide-5-nitrate (IS-5-N, Mono Mack) were examined in a comparative study. The haemodynamic and antianginal properties and the spasmolytic activity on blood vessels
E Malta
Clinical and experimental pharmacology & physiology, 16(11), 829-835 (1989-11-01)
1. The present study has examined the possibility that one or more metabolites of glyceryl trinitrate (GTN) (i.e. glyceryl-1,2- and -1,3-dinitrate and glyceryl-1- and -2-mononitrate) may be responsible for the second phase of the biphasic relaxant curve to GTN in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service