Skip to Content
MilliporeSigma

Skip To

L-045

α-Lactalbumin (from bovine milk)

certified reference material, pkg of 100 mg, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.24
UNSPSC Code:
12352202
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

solid

mol wt

14178 Da

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

pkg of 100 mg

manufacturer/tradename

Cerilliant®

technique(s)

liquid chromatography (LC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

General description

α-Lactalbumin is essential for milk production in the mammary gland. It helps in making lactose, which increases milk volume. It also contains important bioactive peptides and essential amino acids necessary for infant nutrition.
α-Lactalbumin is an important whey protein that regulates the production of lactose in milk and is present in the milk of almost all mammalian species. This full-length protein CRM is suitable as starting material for use in calibrators or controls for a variety of LC-MS/MS testing applications including allergen testing, infant formula testing and dietary or nutritional testing applications.

Features and Benefits

  • Comprehensive identification and characterization of the material have been conducted using various analytical techniques.
  • The certified property value is traceable to national institute of standards and technology (NIST) through calibration verification, employing NIST traceable weights.
  • Balances and NIST traceable weights undergo calibration under ISO 17025 accreditation.
  • Long-term stability assessment has been performed under freezer storage conditions (-10 °C to -25 °C).
  • Stability studies have confirmed a minimum stability duration of 60 months through real-time evaluations.

Preparation Note

  • Weigh the material before usage.
  • Calculate the required amount to be weighed based on the certified property value, considering that the material is hygroscopic.
  • Take precautions to minimize moisture exposure during the weighing process.
  • In quantitative applications, the minimum sample size for intended use is 20 mg.
  • For smaller sample sizes, it is advisable to increase the uncertainty by a factor of 2.

Other Notes

For research and analytical purposes exclusively. This product′s purpose is for testing. THIS PRODUCT IS NOT INTENDED FOR HUMAN OR THERAPEUTIC USE.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L5385L7269L6010
description

certified reference material, pkg of 100 mg, Cerilliant®

description

Type I, ≥85% (PAGE), lyophilized powder

description

≥95% (SDS-PAGE), lyophilized powder

description

Type III, calcium depleted, ≥85% (PAGE), lyophilized powder

grade

certified reference material

grade

-

grade

-

grade

-

format

neat

format

-

format

-

format

-

technique(s)

liquid chromatography (LC): suitable

technique(s)

cell culture | mammalian: suitable, electrophoresis: suitable

technique(s)

cell migration: suitable

technique(s)

indirect ELISA: suitable

mol wt

14178 Da

mol wt

-

mol wt

14,070 Da by calculation

mol wt

14,178 Da by calculation

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

-

feature

-

feature

-

packaging

pkg of 100 mg

packaging

-

packaging

-

packaging

-


Still not finding the right product?

Explore all of our products under α-Lactalbumin (from bovine milk)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters