Skip to Content
MilliporeSigma

H-110

Hydroxyitraconazole

100 μg/mL in methanol, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C35H38Cl2N8O5
CAS Number:
Molecular Weight:
721.63
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Clc1c(ccc(c1)Cl)C3(OC(CO3)COc4ccc(cc4)N5CCN(CC5)c6ccc(cc6)N7C=NN(C7=O)C(C(O)C)C)C[n]2ncnc2

InChI

1S/C35H38Cl2N8O5/c1-24(25(2)46)45-34(47)44(23-40-45)29-6-4-27(5-7-29)41-13-15-42(16-14-41)28-8-10-30(11-9-28)48-18-31-19-49-35(50-31,20-43-22-38-21-39-43)32-12-3-26(36)17-33(32)37/h3-12,17,21-25,31,46H,13-16,18-20H2,1-2H3

InChI key

ISJVOEOJQLKSJU-UHFFFAOYSA-N

General description

Hydroxyitraconazole is an active metabolite of itraconazole, an antifungal drug used for the treatment of fungal infections. Therapeutic drug monitoring of hydroxyitraconazole is utilized in the management of patients receiving itraconazole to ensure therapeutic levels of the drug have been achieved. This Certified Spiking Solution® is suitable as starting material for calibrators, controls, or linearity standards for therapeutic drug monitoring or clinical and diagnostic testing of hydroxyitraconazole in patient blood, serum, or plasma samples by HPLC or LC-MS/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library