Skip to Content
MilliporeSigma

H-062

(±)-Hydroxybupropion-D6 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13D6H12ClNO2
CAS Number:
Molecular Weight:
261.78
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-835-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC(C2(O)C(C)NC(C([2H])([2H])[2H])(C([2H])([2H])[2H])CO2)=CC=C1

InChI

1S/C13H18ClNO2/c1-9-13(16,17-8-12(2,3)15-9)10-5-4-6-11(14)7-10/h4-7,9,15-16H,8H2,1-3H3/i2D3,3D3

InChI key

RCOBKSKAZMVBHT-XERRXZQWSA-N

General description

This stable-labeled internal standard is suitable for quantitation of hydroxybupropion levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. (±)-Hydroxybupropion is a major bupropion metabolie in urine. Bupropion is a pharmaceutical used as an antidepressant and a smoking cessation drug sold under the trade name Wellbutrin® and Zyban®.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Wellbutrin is a registered trademark of GlaxoSmithKline LLC
Zyban is a registered trademark of Everris International B.V.


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library