Skip to Content

F-088

Folic acid-13C5,15N solution

100 μg/mL (Methanol with 0.1 N NaOH), certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C1413C5H19N615NO6
Molecular Weight:
447.35
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (Methanol with 0.1 N NaOH)

format

single component solution

storage temp.

−20°C

SMILES string

[15NH]([13C@@H]([13CH2][13CH2][13C](=O)O)[13C](=O)O)C(=O)c1ccc(cc1)NCc2nc3c(nc(nc3O)N)nc2

InChI

1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/t12-/m0/s1/i5+1,6+1,12+1,13+1,18+1,24+1

InChI key

OVBPIULPVIDEAO-JKYNXJBRSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

WGK

WGK 2

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library