Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-258

Supelco

11-Carboxy cannabinol (11-COOH-CBN) solution

1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Cannabinol-7-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24O4
CAS Number:
Molecular Weight:
340.41
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

description

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol

format

single component solution

storage temp.

-10 to -25°C

General description

11-Carboxy cannabinol (11-COOH-CBN), is a metabolite of CBN found in urine or blood of cannabis users. This certified reference solution is suitable as starting materials in the preparation of calibrators and controls for THC testing by GC/MS or LC/MS in forensic analysis, urine drug testing, or clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service