Skip to Content
MilliporeSigma

C-246

Cannabielsoin (CBE) solution

1 mg/mL (Degassed Methanol), certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (Degassed Methanol)

format

single component solution

storage temp.

-10 to -25°C

SMILES string

O1[C@H]2[C@H]([C@@H](CC[C@@]2(O)C)C(=C)C)c3c1cc(cc3O)CCCCC

InChI

1S/C21H30O3/c1-5-6-7-8-14-11-16(22)19-17(12-14)24-20-18(19)15(13(2)3)9-10-21(20,4)23/h11-12,15,18,20,22-23H,2,5-10H2,1,3-4H3/t15-,18+,20-,21-/m0/s1

InChI key

RBEAVAMWZAJWOI-MTOHEIAKSA-N

General description

Cannabielsoin, also known as CBE, is a non-psychoactive cannabinoid metabolite of CBD. This stable-labeled internal standard is suitable for quantitation of Cannabielsoin levels by LC-MS/MS and GC/MS applications for urine drug testing, pharmaceutical research, clinical toxicology or forensic analysis.
Snap-N-Spike®/Snap-N-Shoot®

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F

flash_point_c

9.7 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library