Skip to Content
MilliporeSigma

Skip To

C-223

7-Hydroxy cannabidiol-D3 (7-OH-CBD-D3) solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H27D3O3
Molecular Weight:
333.48
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

format

single component solution

storage temp.

−20°C

SMILES string

OC1=CC(CCCCC([2H])([2H])[2H])=CC(O)=C1C2C(C(C)=C)CCC(CO)=C2

InChI

1S/C21H30O3/c1-4-5-6-7-15-11-19(23)21(20(24)12-15)18-10-16(13-22)8-9-17(18)14(2)3/h10-12,17-18,22-24H,2,4-9,13H2,1,3H3/i1D3

InChI key

ZELUXPWDPVXUEI-FIBGUPNXSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C-180C-224C-181
format

single component solution

format

single component solution

format

single component solution

format

single component solution

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

manufacturer/tradename

Cerilliant®

manufacturer/tradename

Cerilliant®

manufacturer/tradename

Cerilliant®

manufacturer/tradename

Cerilliant®

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

Snap-N-Spike®/Snap-N-Shoot®


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

51.8 °F

flash_point_c

11 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters