Skip to Content
MilliporeSigma

C-152

Cannabidivarinic Acid (CBDVA) solution

1.0 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H26O4
CAS Number:
Molecular Weight:
330.42
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

cannabis testing

format

single component solution

storage temp.

−70°C

SMILES string

CCCC1=CC(O)=C([C@@H]2C=C(C)CC[C@H]2C(C)=C)C(O)=C1C(O)=O

InChI

1S/C20H26O4/c1-5-6-13-10-16(21)18(19(22)17(13)20(23)24)15-9-12(4)7-8-14(15)11(2)3/h9-10,14-15,21-22H,2,5-8H2,1,3-4H3,(H,23,24)/t14?,15-/m1/s1

InChI key

CZXWOKHVLNYAHI-YSSOQSIOSA-N

General description

This certified Snap-N-Spike® solution is suitable for CBDVA testing methods by GC/MS, LC/MS or HPLC for applications in testing of Cannabis potency or impurity profiling and pharmaceutical research. Cerilliant solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK#s for Canadian customers.

Cannabidivarinic acid (CBDVA) is a non-psychoactive cannabinoid found in Cannabis that reportedly has anti-inflammatory properties.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:

  • Serum analysis to identify and quantify 18 phytocannabinoids by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Quantitative analysis of 17 cannabinoids in cannabis and hemp samples by LC-MS/MS following their liquid-solid extraction following AOAC and ASTM guidelines for cannabis and hemp matrices
  • Liquid chromatographic (LC)-diode array detection (DAD) based separation and measurement of 20 cannabinoids from a variety of cannabis samples
  • Untargeted analysis of extracts from cultivars of 23 Cannabis sativa L plant materials by gas chromatography–time-of-flight/mass spectrometry (GC-TOF/MS) and liquid chromatography quadrupole time-of-flight tandem mass spectrometry (LC-QTOF MS/MS)

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library