Skip to Content
MilliporeSigma

C-122

2C-B-FLY hydrochloride solution

1 mg/mL in acetonitrile: water (9:1), ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H14BrNO2 · HCl
CAS Number:
Molecular Weight:
320.61
NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in acetonitrile: water (9:1)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

BrC1=C2C(OC=C2)=C(CCN)C3=C1OC=C3.Cl

InChI

1S/C12H10BrNO2.ClH/c13-10-9-3-6-15-11(9)7(1-4-14)8-2-5-16-12(8)10;/h2-3,5-6H,1,4,14H2;1H

InChI key

COTFOIBRWRDLCY-UHFFFAOYSA-N

General description

2C-B-FLY is a hallucinogen related to the 2C amine family of illicit drugs such as 2C-B, 2C-I, 2C-E, and 2C-C. Psychedelic effects of this designer drug usually last from 6-8 hours with up to 12 hours observed for larger doses. This certified solution standard is suitable for 2C amine testing methods by GC/MS or LC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F

flash_point_c

2.0 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library