Skip to Content
MilliporeSigma
All Photos(1)

Documents

B-906

Supelco

Benzyl piperazine dihydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2 · (HCl)2
CAS Number:
Molecular Weight:
249.18
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.Cl.C1CN(CCN1)Cc2ccccc2

InChI

1S/C11H16N2.2ClH/c1-2-4-11(5-3-1)10-13-8-6-12-7-9-13;;/h1-5,12H,6-10H2;2*1H

InChI key

BBUJLUKPBBBXMU-UHFFFAOYSA-N

General description

A certified solution standard suitable for LC/MS or GC/MS amphetamine testing methods in clinical toxicology, forensic analysis, or urine drug testing. Benzyl piperazine, or BZP, is an amphetamine analog used illicitly for its euphoriant and stimulant effects.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1-benzylpiperazine (BZP) abuse amongst attendees of the Drug Treatment Centre Board.
S McNamara
Irish medical journal, 102(6), 191-191 (2009-09-03)
Amber J Dickson et al.
Journal of analytical toxicology, 34(8), 464-469 (2010-01-01)
Historically, ecstasy tablets contained 3,4-methylenedioxymethamphetamine (MDMA) as the psychoactive component. In recent years, the Drug Enforcement Administration (DEA) and other law enforcement agencies have seized ecstasy tablets that are comprised of psychoactive drugs or drug mixtures other than MDMA. Many
Poison in party pills is too much to swallow.
Mike Cole
Nature, 474(7351), 253-253 (2011-06-17)
David M Wood et al.
Therapeutic drug monitoring, 34(4), 363-367 (2012-06-08)
Since the turn of the 21st century, there has been an increase in the availability and use of novel psychoactive substances (also known as "legal highs") across Europe. Currently, there is limited information available on the potential acute toxicity (harms)
Chris Wilkins et al.
Drug and alcohol review, 27(6), 633-639 (2009-04-21)
A large legal market for party pills containing benzylpiperazine (BZP) and trifluoromethylphenylpiperazine (TFMPP) developed in New Zealand after 2004. The use of these party pills has been associated with adverse health effects. The purpose of this paper was to assess

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service