Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C20H22BrN3O · HCl
Molecular Weight:
436.77
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
grade
certified reference material
Quality Segment
packaging
ampule of 1 mL ((as free base))
manufacturer/tradename
Cerilliant®
drug control
kontrollierte Droge in Deutschland
concentration
1 mg/mL in methanol
format
single component solution
storage temp.
-10 to -25°C
General description
Brorphine is a piperidine-based opioid analgesic sold and abused as designer drug. A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls in brorphine LC/MS or GC/MS testing methods for clinical toxicology, urine drug testing or forensic analysis applications.
Snap-N-Spike®/Snap-N-Shoot®
Legal Information
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.
English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.
English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
WGK
WGK 2
Flash Point (°F)
49.5 °F
Flash Point (°C)
9.7 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


