Select a Size
About This Item
grade
certified reference material
Quality Segment
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol
format
single component solution
storage temp.
−20°C
SMILES string
O[C@@H]1[C@@H](C(O)=O)O[C@@H](OC2=C(O3)C([C@]([C@@H]3[C@](CC4)(OC)C(C(O)(C)C(C)(C)C)C5)(CCN6C([2H])([2H])C7C([2H])([2H])C7)[C@@]54[C@H]6C8)=C8C=C2)[C@H](O)[C@H]1O
InChI
1S/C35H49NO10/c1-31(2,3)32(4,42)20-15-33-10-11-35(20,43-5)30-34(33)12-13-36(16-17-6-7-17)21(33)14-18-8-9-19(26(46-30)22(18)34)44-29-25(39)23(37)24(38)27(45-29)28(40)41/h8-9,17,20-21,23-25,27,29-30,37-39,42H,6-7,10-16H2,1-5H3,(H,40,41)/t20?,21-,23+,24+,25-,27+,29-,30-,32?,33-,34+,35-/m1/s1/i6D2,16D2/t17?,20?,21-,23+,24+,25-,27+,29-,30-,32?,33-,34+,35-
InChI key
CZULHKGIAJASAA-QTQVLIFGSA-N
General description
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


