Skip to Content
MilliporeSigma

B-051

Buphedrone Ephedrine Metabolite-D3 hydrochloride solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H14D3NO · HCl
Molecular Weight:
218.74
UNSPSC Code:
41116107
MDL number:
NACRES:
NA.24
EC Number:
200-659-6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

OC(C(CC)NC([2H])([2H])[2H])C1=CC=CC=C1.[H]Cl

InChI

1S/C11H17NO.ClH/c1-3-10(12-2)11(13)9-7-5-4-6-8-9;/h4-8,10-13H,3H2,1-2H3;1H/i2D3;

InChI key

DBIYVUUUZVZMQD-MUTAZJQDSA-N

General description

A stable-labeled internal standard suitable for quantitation of Buphedrone Ephedrine Metabolite levels in urine or other matrices by LC/MS or GC/MS for testing applications in clinical toxicology, forensic analysis, or urine drug testing. This bath salt cathinone analog is a primary urinary metabolite of Buphedrone. Buphedrone, also known as MAPB, is a synthetic cathinone and stimulant widely found in numerous bath salt blends across the US, Europe, and Asia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library