Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A-924

Supelco

7-Aminoclonazepam-d4 solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D4H8ClN3O
CAS Number:
Molecular Weight:
289.75
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

O=C1NC2=CC=C(N)C=C2C(C3=C([2H])C([2H])=C([2H])C([2H])=C3Cl)=NC1

InChI

1S/C15H12ClN3O/c16-12-4-2-1-3-10(12)15-11-7-9(17)5-6-13(11)19-14(20)8-18-15/h1-7H,8,17H2,(H,19,20)/i1D,2D,3D,4D

InChI key

HEFRPWRJTGLSSV-RHQRLBAQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An internal standard suitable for use in LC/MS or GC/MS testing methods of 7-aminoclonazepam for clinical toxicology, pain prescription monitoring, forensic analysis, or isotope dilution methods. 7-Aminoclonazepam is a major metabolite of clonazepam, a potent, long-acting benzodiazepine marketed as Klonopin® in the US and Rivotril in many other countries.

Application


  • Intralipid as a matrix additive for evaluating hyperlipidemic postmortem blood: This study explores the utility of specific matrix additives for postmortem toxicological evaluations, which can be instrumental when standardizing methods involving metabolites like 7-Aminoclonazepam-d₄ in forensic toxicology analysis (Elenstål et al., 2023).

  • Comparison of Two Immunoassay Screening Methods and a LC-MS/MS in Detecting Traditional and Designer Benzodiazepines in Urine: This investigation into immunoassay and mass spectrometry methods for drug screening underscores the utility of deuterated standards such as 7-Aminoclonazepam-d₄ for ensuring the precision of pharmacological research (Rossi et al., 2021).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Klonopin is a registered trademark of Hoffmann-La Roche Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service