Skip to Content
MilliporeSigma

A-061

Multicomponent Alcohol Mix-4000

4000 μg/mL each component in H2O, ampule of 1.2 mL, certified reference material, Cerilliant®

Synonym(s):

Multi-component Alcohol Mix

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Acetone , Ethanol , Isopropanol , Methanol

packaging

ampule of 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

4000 μg/mL each component in H2O

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

multi-component solution

storage temp.

2-8°C

InChI

1S/C15H13N3O/c16-11-6-7-13-12(8-11)15(17-9-14(19)18-13)10-4-2-1-3-5-10/h1-8H,9,16H2,(H,18,19)/i1D,2D,3D,4D,5D

InChI key

OYOUQHVDCKOOAL-RALIUCGRSA-N

General description

A multicomponent calibration kit containing acetone, ethanol, isopropanol, and methanol at a concentration of 4000 μg/mL. This kit is suitable for calibration of blood alcohol testing methods using GC Headspace.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library