Skip to Content
MilliporeSigma
All Photos(1)

Documents

A-057

Supelco

Multicomponent Alcohol Mix 500

500 μg/mL each component in H2O, ampule of 1.2 mL, certified reference material, Cerilliant®

Synonym(s):

Multi-component Alcohol Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Acetone
Ethanol
Isopropanol
Methanol

packaging

ampule of 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

500 μg/mL each component in H2O

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

multi-component solution

storage temp.

2-8°C

InChI

1S/C15H13N3O/c16-11-6-7-13-12(8-11)15(17-9-14(19)18-13)10-4-2-1-3-5-10/h1-8H,9,16H2,(H,18,19)/i1D,2D,3D,4D,5D

InChI key

OYOUQHVDCKOOAL-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A multicomponent calibration kit containing acetone, ethanol, isopropanol, and methanol at a concentration of 500 μg/mL. This kit is suitable for calibration of blood alcohol testing methods using GC Headspace.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service