Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

850151P

Avanti

18:1 PI(4)P

Avanti Research - A Croda Brand 850151P, powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phospho-(1′-myo-inositol-4′-phosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H90N2O16P2
CAS Number:
Molecular Weight:
977.15
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850151P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850151P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850151P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](OP([O-])(O)=O)[C@@H](O)[C@H]1O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+].[NH4+]

InChI

1S/C45H84O16P2.2H3N/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(46)57-35-37(59-39(47)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)36-58-63(55,56)61-45-42(50)40(48)44(41(49)43(45)51)60-62(52,53)54;;/h17-20,37,40-45,48-51H,3-16,21-36H2,1-2H3,(H,55,5

InChI key

XDKQBHORSIMQSB-QVUMANCCSA-N

General description

18:1 PI(5)P (1,2-dioleoyl-sn-glycero-3-phospho-(1′-myo-inositol-4′-phosphate) is an ammonium salt of modified lipid which bears a double at the 9th carbon of dioleoyl group. Phosphatidylinositol 4-phosphate (PI-4-P) is an abundant phosphoinositide species present in the plasma membrane.
Inositol phospholipid species are membrane-bound signaling molecules that have been implicated in almost all aspects of cellular physiology, including cellular growth, metabolism, proliferation, and survival.

Application

18:1 PI(5)P (1,2-dioleoyl-sn-glycero-3-phospho-(1′-myo-inositol-4′-phosphate) (ammonium salt)) has been used in protein−lipid overlay assay. It may be used in small unilamellar vesicles for liposome flotation assays. It may also be used in different lipid binding specificity studies.

Biochem/physiol Actions

Phosphatidylinositol 4-phosphate (PI-4-P) is an important factor, that is required for membrane traffic, polarized growth and cytoskeleton organization.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850151P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850151P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vikram Ghugtyal et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(28), 8644-8649 (2015-07-01)
The phospholipid phosphatidylinositol-4-phosphate [PI(4)P], generated at the Golgi and plasma membrane, has been implicated in many processes, including membrane traffic, yet its role in cell morphology changes, such as the budding to filamentous growth transition, is unknown. We show that
Carolyn M Shirey et al.
Analytical biochemistry, 516, 9-12 (2016-10-28)
To reduce costs of lipid-binding assays, allow for multiple lipids to be screened for protein binding simultaneously, and to make lipid binding more user friendly, lipids have been dotted onto membranes to investigate lipid-protein interactions. These assays are similar to
Chao-Yuan Yu et al.
Plant physiology, 183(1), 221-235 (2020-03-25)
Phosphoinositides function as lipid signals in plant development and stress tolerance by binding with partner proteins. We previously reported that Arabidopsis (Arabidopsis thaliana) phosphoinositide-specific phospholipase C2 functions in the endoplasmic reticulum (ER) stress response. However, the underlying molecular mechanisms of
Mark R Lundquist et al.
Molecular cell, 70(3), 531-544 (2018-05-05)
While the majority of phosphatidylinositol-4, 5-bisphosphate (PI-4, 5-P2) in mammalian cells is generated by the conversion of phosphatidylinositol-4-phosphate (PI-4-P) to PI-4, 5-P2, a small fraction can be made by phosphorylating phosphatidylinositol-5-phosphate (PI-5-P). The physiological relevance of this second pathway is
Lee Dolat et al.
The Journal of cell biology, 214(5), 517-527 (2016-08-24)
Macropinocytosis, the internalization of extracellular fluid and material by plasma membrane ruffles, is critical for antigen presentation, cell metabolism, and signaling. Macropinosomes mature through homotypic and heterotypic fusion with endosomes and ultimately merge with lysosomes. The molecular underpinnings of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service