Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

845875P

Avanti

18:1 Lyso PC

Avanti Research - A Croda Brand

Synonym(s):

1--(9Z-octadecenoyl)-sn-glycero-3-phosphocholine; PC(18:1(9Z)/0:0); 110688

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H52NO7P
CAS Number:
Molecular Weight:
521.67
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1-oleoyl-2-hydroxy-sn-glycero-3-phosphocholine

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 2 × 100 mg (845875P-200mg)
pkg of 5 × 100 mg (845875P-500mg)
pkg of 1 × 25 mg (845875P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C26H52NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-26(29)32-23-25(28)24-34-35(30,31)33-22-21-27(2,3)4/h12-13,25,28H,5-11,14-24H2,1-4H3/b13-12-/t25-/m1/s1

InChI key

YAMUFBLWGFFICM-PTGWMXDISA-N

General description

Lysophosphatidylcholine (LPC) is a lysophospholipid mediator. This bioactive proinflammatory lipid is produced by pathological activities. It is one of the important constituent of oxidized low-density lipoprotein (Ox-LDL).

Application

18:1 Lyso PC is suitable for use as a lysophospholipid mediator to compare the effect of sphingosylphosphorylcholine (SPC). It has been used as a supplement in Yeast nitrogen base (YNB) medium to rescue choline auxotrophy.

Biochem/physiol Actions

Lysophosphatidylcholine (LPC) participates as a key factor in the atherogenic activity of oxidized low-density lipoprotein (Ox-LDL). It alters the roles in endothelial cells, smooth muscle cells, monocytes, macrophages, T-cells, thereby involving in atherosclerosis and inflammatory diseases. LPC activates protein kinase C, extracellular-signal-regulated kinases, protein tyrosine kinases, and Ca(2+), that are second messengers.

Packaging

5 mL Clear Glass Sealed Ampule (845875P-200mg)
5 mL Clear Glass Sealed Ampule (845875P-25mg)
5 mL Clear Glass Sealed Ampule (845875P-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysophospholipid receptors: signalling, pharmacology and regulation by lysophospholipid metabolism
zu HDM and Jakobs KH
Biochimica et Biophysica Acta - Biomembranes, 1768(4), 923-940 (2007)
Overproduction of phospholipids by the Kennedy Pathway Leads to Hypervirulence in Candida albicans
Tams RN, et al.
Frontiers in Microbiology, 10(2), 86-86 (2019)
Regulation of ryanodine receptors by sphingosylphosphorylcholine: Involvement of both calmodulin-dependent and-independent mechanisms
Kovacs E, et al.
Biochemical and biophysical research communications, 401(2), 281-286 (2010)
Jacinto López-Sagaseta et al.
The EMBO journal, 31(8), 2047-2059 (2012-03-08)
Invariant Natural Killer T (iNKT) cells use highly restricted αβ T cell receptors (TCRs) to probe the repertoire of lipids presented by CD1d molecules. Here, we describe our studies of lysophosphatidylcholine (LPC) presentation by human CD1d and its recognition by
Danielle M Charron et al.
Langmuir : the ACS journal of surfaces and colloids, 36(19), 5385-5393 (2020-04-28)
Porphyrin aggregates have attractive photophysical properties for phototherapy and optical imaging, including quenched photosensitization, efficient photothermal conversion, and unique absorption spectra. Although hydrophobic porphyrin photosensitizers have long been encapsulated into liposomes for drug delivery, little is known about the membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service