Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

600200

Avanti

10mL Pre-Scored Ampule, Clear

Avanti Research - A Croda Brand

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24122000
NACRES:
NA.28

packaging

pkg of 1 × 1 ea

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

ambient

storage temp.

room temp

Packaging

10mL Clear Ampule

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service