Key Documents
Z766852
Microflex NeoPro® EC powder-free chloroprene gloves
size M
Synonym(s):
disposable glove
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
(chloroprene gloves)
green
size
M
description
ambidexterous
textured fingers
feature
CE compliant
disposable
latex free
powder-free
packaging
pack of 50 ea (boxes of 50)
manufacturer/tradename
Microflex NEC-288-M
L
11.8 in. , cuff
thickness
6.3 mil , palm
8.3 mil , fingers
General description
Featureing an extended cuff, NeoPro® EC is the longest and thickest non-latex glove produced by Microflex. It uses a polychloroprene formulation to provide a secure grip along with exceptional protection from biological and chemical hazards inwet or dry conditions.
Legal Information
NeoPro is a registered trademark of CIT Healthcare LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)