Key Documents
Z755575
Stacking platform for Mini BlotBoy
pkg of 1 ea
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
packaging
pkg of 1 ea
manufacturer/tradename
Benchmark Scientific B3D-STACK
General description
Stacking platform, small 10.5x7.5 inches with flat mat and giving 3.0 inche separation of platforms
Legal Information
Mini BlotBoy is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)