Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z743892

ART® SoftFit L Reload System

Pure 1000L, size 1000 μL, hinged rack, sterile

Synonym(s):

aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

sterility

sterile

feature

Pure 1000L
hinged rack

packaging

pkg of 4X768 ea

manufacturer/tradename

Molecular BioProducts 3781-HR

technique(s)

PCR: suitable

size

1000 μL

Looking for similar products? Visit Product Comparison Guide

General description

“SoftFit-L” pipette tips can be used to achieve the accuracy and precision you require for your pipette samples consistently.

Application

“ART”SoftFit L Reload System is suitable for:,·      sensitive applications and samples (cell culture, PCR, nucleic acid work, etc.),·      volatile or corrosive samples

Features and Benefits

  • Hinged rack enables effortless one-hand access to tips.
  • Color-coded inserts facilitate swift identification of tip volume.
  • Precise fit is guaranteed by the cylindrical collar and positive-stop design, leading to reduced attachment and ejection forces.
  • Tips are certified to be free of RNase, DNase, DNA, ATP, and endotoxin contamination.
  • For traceability, tip inserts are labeled with lot and product part numbers.
  • Effectively block aerosols from contaminating the pipette and prevent cross-contamination of samples.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service