Key Documents
Z743214
Julabo® bath accessories
installation cooing coil, for CORIO C and CD ranges, pack of 1 ea
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9970101
General description
Add-on cooling coil (9 mm outer diameter) for counter-cooling with cooling water, can be used for tempering tasks close to the ambient temperature. A hose with an inner diameter of 10 mm is required. For use with Julabo® baths/circulators.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.