Key Documents
Z743180
Julabo® bath covers
lift-up gable cover to fit CORIO CD-B33
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9970257
General description
The hinged stainless steel bath lid reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the hood prevents contamination of the samples in the bath.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)