Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z742917

CITRANOX® acid detergent, biodegradable

pack of 947 mL

Synonym(s):

biodegradable cleaner, low foaming acid detergent, metal oxide remover, phosphate-free cleaner, ultrasonic cleaner detergent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.34

packaging

pack of 947 mL

manufacturer/tradename

Alconox, Inc. 1832

Looking for similar products? Visit Product Comparison Guide

Legal Information

CITRANOX is a registered trademark of Alconox, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service