Key Documents
Z742901
Kern ABT analytical balances
Kern ABT 100-5NM, weighing capacity 101 g, accuracy: 0.01 g, precision: ±0.15 g
Synonym(s):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
About This Item
UNSPSC Code:
41111502
NACRES:
NB.71
AC/DC input
100 - 240 V AC, universal plug set
packaging
pack of 1 ea
manufacturer/tradename
Kern ABT 100-5NM
weighing capacity measuring range
101 g, accuracy: 0.01 g, precision: ±0.15 g
General description
The Kern ABT analytical balances are the premium balance model with a high-quality single cell weighing system. It has a unique dosage aid which has a high-stability mode and other filter settings. The simple recipe weighing and documenting is provided with a combined tare/print function, and the simple adding function has auto-tare and automatic subsequent data output after each weighing procedure.