Key Documents
Z742349
2MAG Asteroid extra strength stirring bar
diam. × L 39 mm × 70 mm
Synonym(s):
magentic flea, stirbar
About This Item
UNSPSC Code:
41103800
NACRES:
NB.72
material
PTFE cover
interior (SmCo)
triangular
packaging
pack of 1 ea
manufacturer/tradename
2mag 44070
diam. × L
39 mm × 70 mm
weight
0.3 kg
General description
New developed, high-effective stirring bar, ideal for large mixing volumes, viscous media, stirring over wide distances or for disadvantageous vessel bottoms.
More than 4x higher torque transmissible, unbeatable increased mixing effect in comparison to conventional stirring bars, high energetic and long-term stable magnetic field, no demagnetization effect, concave design for a minimum of bearing surface and reliable rotation, triangular shape for powerful shear force, better mixing effect at lower rotation speed.
300 percent less PTFE abrasion than comparable round shape stirring bars, significantly higher rotation speed reachable, high quality, even and sealed PTFE cover, FDA-conform, steam sterilizable at 121 degrees C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)