Documents
Z741984
Büchi® Rotavapor R-100
cold trap, AC/DC input 100 AC, US 3-pin plug
Synonym(s):
Rotavapor
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
Joint
joint SJ24/40
manufacturer/tradename
Büchi 11100C202
packaging
1 ea of
General description
The rotary evaporator R-100 is a high-quality, entry level Rotavapor to meet the essential needs in classical laboratory applications. It is capable of a manually adjustable rotation speed of 20 to 280 rpm and equipped with an action jack lift for manual lifting of the evaporating flask. Included is the selected condenser, 1 L receiving and evaporating flasks, and a vapor duct. All accessories are included for out of the box usage.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG