Key Documents
Z740943
Sigma-Aldrich
Aldrich® Essentials culture tube with screw cap and PTFE/rubber liner
capacity 10 mL, O.D. × L 15 mm × 125 mm, round bottom
About This Item
UNSPSC Code:
41121706
NACRES:
NB.11
material
borosilicate glass tube
round bottom
screw top cap
screw top neck
transparent tube
packaging
pack of 72 ea
manufacturer/tradename
Sigma-Aldrich
O.D. × L
15 mm × 125 mm
capacity
10 mL
General description
Clear borosilicate glass, round bottom culture tubes with thread top and black phenolic cap. Cap has bonded PTFE faced rubber liner.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)