Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z740658

Sigma-Aldrich

Aldrich® Essentials Erlenmeyer flask, narrow mouth

capacity 1000 mL, stopper joint: 9, volume 250-1000 mL, accuracy: 50 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

material

borosilicate glass flask
flat bottom

Joint

joint: stopper9

sterility

non-sterile

feature

without cap

packaging

pkg of 6 ea

volume measuring range

250-1000 mL, accuracy: 50 mL

capacity

1000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Clear borosilicate glass, standard wall Erlenmeyer flasks for standard laboratory use. Narrow mouth opening with beaded rim. Reference graduations and marking spot in white. ASTM E 1404, DIN 12380.

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service