Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z699020

Orange serum Broth

pkg of 50 ea

Synonym(s):

orange serum agar, tryptone soy broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111700
NACRES:
NB.79

packaging

ampule of 50 × 2 mL
pkg of 50 ea

manufacturer/tradename

Maine Mfg 10496104

application(s)

food and beverages

General description

Organisms known to grow in single strength and concentrated juices are lactic acid and acetic acid bacteria and yeast. Lactobacilli, Leuconostoc and yeast have all been identified as spoilage organisms by numerous authors. Orange serum at pH 5.4 to 5.6 has been reported to yield maximum counts of all types of spoilage organisms in mixed cultures and in single culture comparison tests.

The low pH of the test solution usually prohibits the development of other microorganisms which cannot survive low pH conditions. Therefore developing colonies are presumed to be problematic organisms.

Formulation.
Per liter of water, adjusted to pH 5.6 at 25oC
  • Orange serum100ml
  • Yeast extract3.0g
  • Tryptone10.0g
  • Dextrose4.0g
  • Dipotassium phosphate2.5g

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service